Learn More
Hampton Research NATRIX NO48 185ML
SDP
Catalog No. 501751
Encompass
HR2-918-48/Natrix #48 (D12), 185 ml - 0.2 M Ammonium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M TRIS hydrochloride pH 8.5, 30 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.
Catalog No. 50-175-1 Supplier Hampton Research Supplier No. HR291848
HR2-918-48/Natrix #48 (D12), 185 ml - 0.2 M Ammonium chloride, 0.01 M Calcium chloride dihydrate, 0.05 M TRIS hydrochloride pH 8.5, 30 % w/v Polyethylene glycol 4,000. Please note items from this supplier are non-returnable.