Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ BC/GN Panel
SDP

Catalog No. 22156109 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
12 X 0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-109 12 X 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-109 Supplier ZeptoMetrix Supplier No. NATBCGNNNS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to evaluate the performance of nucleic acid tests for the determination of the presence of bacterial nucleic acids

  • Formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Product Type BC/GN Panel
For Use With (Application) Evaluate performance of nucleic acid tests for determination of presence of bacterial nucleic acids
Quantity 12 X 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.