Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ BC/GP Panel
SDP

Catalog No. 22156111 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
0.75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-111 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-111 Supplier ZeptoMetrix Supplier No. NATSPNNNS
Add to Cart
Add to Cart

Formulated with purified, chemically modified intact bacterial particles for QC validation

  • Contains 0.75mL vial of bacterial NATtrol™ target S. pneumoniae, Strain: Z022
  • Supplied in purified protein matrix that is identical to the composition of true clinical specimen

Contains intact organisms and should be run in a manner identical to that used for clinical specimens

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Product Type BC/GP Panel
For Use With (Application) Evaluate performance of nucleic acid tests for determination of presence of bacterial nucleic acids
Quantity 0.75 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.