Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ BC/GP Panel
GSA_VASDP

Catalog No. 22156884 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
11 x 0.7 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-884 11 x 0.7 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-884 Supplier ZeptoMetrix Supplier No. NATBC/GPNNS
Add to Cart
Add to Cart

Formulated with purified, chemically modified intact bacterial particles for QC validation

  • Contains 11 x 0.75mL vials of bacterial NATtrol™ targets
  • Supplied in purified protein matrix that is identical to the composition of true clinical specimen
    • Contains 11 x 0.75mL vials of bacterial NATtrol™ targets
    • Supplied in purified protein matrix that is identical to the composition of true clinical specimen
    • S. aureus (MRSA), Strain: COL
    • S. epidermidis (MSSE), Strain: HER1292
    • S. lugdunensis, Strain: Z097
    • S. pneumoniae, Strain: Z022
    • S. aginosus/intermedius, Strain: CL-CS 56
    • S. agalactiae, Strain: Z019
    • S. pyogenes, Strain: Z018
    • E. faecalis, Strain: VRE
    • E. faecium, Strain: Van A MCW-CS - 166
    • L. monocytogenes, Strain: N/A
    • Negative, Strain: N/A

    Contains intact organisms and should be run in a manner identical to that used for clinical specimens

    Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Verification Panel
For Use With (Application) Determination of presence of bacterial Nucleic Acids
Quantity 11 x 0.7 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.