Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ BK Virus Linearity Panel
SDP

Catalog No. 22156710 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-710 6 x 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-710 Supplier ZeptoMetrix Supplier No. NATBKLIN
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Contains 6 x 0.25mL vials of BK Virus NATtrol™ at concentrations 0 to 100,000,000 copies/mL
  • Supplied in purified protein matrix identical to composition of true clinical specimen

Contains intact virus and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 0 to 100000000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Linearity Panel
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of Human BK virus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 0.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.