Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ BV Negative Control
SDP

Catalog No. 22158099 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-099 6 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-099 Supplier ZeptoMetrix Supplier No. NATBVNEGBD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to evaluate the performance of nucleic acid tests. ZeptoMetrix NATtrol™ BV Negative Control is used for quality control of clinical assays and training of laboratory personnel.

  • Formulated with purified, intact bacterial and fungal cells that have been chemically modified to render them non-infectious
  • Contains 6 x 0.5 mL vials of NATtrol™ BV Positive Control or NATtrol™ BV Negative Control
  • Evaluates the performance of nucleic acid tests for determination of the presence of BV DNA markers
  • Positive or Negative BV markers are represented depending on marker combination used
Controls:
  • NATBVPOS-BD (Positive) - Organism: L. jensenii, G. vaginalis, A. vaginae, BVAB2; Strain: Z264, Z247, Z242, recombinant
  • NATBVNEGBD (Negative) - Organism: L. crispatus, G. vaginalis; Strain: Z246, Z247

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Negative Control
For Use With (Application) Determination of the presence of BV DNA markers
Quantity 6 x 0.5 mL

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.