Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Coxsackievirus B3
SDP

Catalog No. 22157776 Shop All ZeptoMetrix Corporation Products
Click to view available options
Quantity:
1 mL

Catalog No. 22-157-776

Supplier: ZeptoMetrix NATCXVB3ST

Only null left
Add to Cart
Add to Cart

Qualitative molecular control

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Virus Stock
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of Coxsackievirus RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.