Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol CT/NG/TV/MG Positive Control Swab
SDP

Catalog No. 22042946 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
4 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-042-946 4 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-042-946 Supplier ZeptoMetrix Supplier No. NATSTISW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains purified, intact microorganisms that have been chemically modified to render them non-infectious

  • Includes positive and negative external run controls intended to be used with molecular assays that detect nucleic acids from Adenovirus F40/41, Astrovirus, Blastocystis spp., Campylobacter spp., Clostridioides difficile (tcdA/tcdB), Cryptosporidium spp., Cyclospora cayetanensis, Dientamoeba fragilis, Entamoeba histolytica, Enterotoxigenic E. coli (ETEC) LT/ST, Giardia lamblia, Microsporidia, Norovirus GI/GII, Plesiomonas shigelloides, Rotavirus A, Salmonella spp., Sapovirus I/II/IV/V, Shiga-like toxin-producing E. coli (STEC) stx1, Shiga-like toxin-producing E. coli (STEC) stx2, Shigella/Enteroinvasive E. coli (EIEC), Strongyloides stercoralis, Vibrio spp., Vibrio cholerae, and Yersinia enterocolitica
  • The positive controls contain purified, intact microorganisms and viruses that have been chemically modified to render them non-infectious and refrigerator stable and are formulated in a proprietary matrix
  • The negative control is formulated in the absence of any microorganisms or viruses and contains proprietary matrix only
  • The controls can be used to monitor the process steps of extraction, amplification and detection
  • The controls can also be used for training and evaluation of operator variation

Specifications

Type Control Swab
Quantity 4 Ea.
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.