Learn More
ZeptoMetrix NATtrol™ Cytomegalovirus AD-169 External Run Controls
Shop All ZeptoMetrix Corporation Products

Description
- Each control pack contains 6 x 1.0mL vials of CMV NATtrol™ at concentrations 5,000 to 50,000 copies/mL
- Supplied in purified protein matrix identical to composition of true clinical specimen
- Designed to evaluate the performance of nucleic acid tests for determination of the presence of CMV DNA
Validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Concentration | 5000 copies/mL |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | NATtrol External Run Control |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of CMV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.