Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Cytomegalovirus Linearity Panel, AD-169 Strain
SDP

Catalog No. 22156719 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-719 6 x 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-719 Supplier ZeptoMetrix Supplier No. NATCMVLIN
Add to Cart
Add to Cart

Designed to evaluate the performance of nucleic acid tests for determination of the presence of CMV DNA

  • Formulated with purified, intact virus particles that are chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Contains 6 x 0.25mL panel members

  • Panel Members:
  • CMV 5E6: 5,000,000copies/mL
  • CMV 5E5: 500,000copies/mL
  • CMV 5E4: 50,000copies/mL
  • CMV 5E3: 5,000copies/mL
  • CMV 5E2: 500copies/mL
  • Negative: 0

NATCMV-LIN contains intact organisms and should be run in a manner identical to that used for clinical specimens

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 0 to 5000000 copies/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Linearity Panel
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of CMV DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 0.25 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.