Learn More
Specifications
Specifications
| Certifications/Compliance | ISO, 13485 |
| Content And Storage | 2°C to 8°C |
| Form | Solution |
| Media Type | Vial |
| Product Type | NATtrol Virus Stock |
| Formulation | Formulated with purified, intact virus particles |
| For Use With (Application) | Determination of presence of Echovirus RNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel |
| Quantity | 1 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
