Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Echovirus 6
SDP

Catalog No. 22157780 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-157-780 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Overexposure will cause possible irritation with the eyes and skin. This is toxic to fish due to containing 0.09% sodium azide.
Catalog No. 22-157-780 Supplier ZeptoMetrix Supplier No. NATECHO6ST
Add to Cart
Add to Cart
Overexposure will cause possible irritation with the eyes and skin. This is toxic to fish due to containing 0.09% sodium azide.

NATtrol™ Echovirus Type 6 Stock (NATECHO6-ST) is formulated with purified, intact virus particles that have been chemically modified

Contains intact virus and should be run in a manner identical to that used for clinical specimens

  • Appearance: clear, pale amber color
  • Noninfectious and refrigerator stable
  • Store at 2° to 8° C
  • Purified, intact organisms
  • pH is 7.5 +/- 0.5
  • Quantity 1 mL

Designed to evaluate the performance of nucleic acid tests for determination of the presence of Echovirus RNA, and can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Certifications

ISO 13485

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Virus Stock
Formulation Formulated with purified, intact virus particles
For Use With (Application) Development of diagnostic tests, training of laboratory personnel and validation of clinical assays
Quantity 1 mL

To aviod cross contamination, use separate pipette tips for all reagents. Extract Echovirus RNA prior to use in downstream assays. Use Universal precautions when handling this product. This product is stable, just avoid contact with metals (aluminum, lead, copper, zinc, mercury) and acids.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.