Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ EV Negative Control
SDP

Catalog No. 22158040 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-040 6 x 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-040 Supplier ZeptoMetrix Supplier No. NATEVNEG6MC
Add to Cart
Add to Cart

Designed for use in evaluating and monitoring the performance of nucleic acid tests for determination of the presence of EV nucleic acids

  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Can be used for verification of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Content And Storage 2°C to 8°C
Quantity 6 x 0.2 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.