Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol Flu/RSV/SARS-CoV-2 Negative Control Swab
SDP

Catalog No. 21279115 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
12/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-279-115 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-279-115 Supplier ZeptoMetrix Supplier No. NATFRCNEGSWIVD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures.

NATtrol™ Flu/RSV/SARS-CoV-2 Negative Control Swab contains human A-549 cells formulated in a proprietary matrix and dried on swabs. NATtrol™ Flu/RSV/SARS-CoV-2 Negative Control Swab is a full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures

Specifications

Detectable Analytes SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV)
Format Swab
Sample Type Swab
Type Negative Control Swab
Quantity 12/Cs.
Form Freeze-dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.