Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol Flu/RSV/SARS-CoV-2 Positive Control Swab
SDP

Catalog No. 21279116 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
12/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-279-116 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 21-279-116 Supplier ZeptoMetrix Supplier No. NATFRCSWIVD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures.

NATtrol™ Flu/RSV/SARS-CoV-2 Positive Control Swab contains purified, intact viral particles that have been chemically modified to render them non-infectious and human A-549 cells. Each swab contains Influenza A (A/Brisbane/10/07), Influenza B (B/Florida/02/06), RSV B (CH93(18)-18), SARS-CoV-2 (USA-WA1/2020)1 and A-549 cells. The analytes are formulated in a proprietary matrix and dried on swabs. NATtrol™ Flu/RSV/SARS-CoV-2 Positive Control Swab is a full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures.

Specifications

Detectable Analytes SARS-CoV-2, Influenza A and B, Respiratory Syncytial Virus (RSV)
Format Swab
Sample Type Swab
Type Positive Control Swab
Quantity 12/Cs.
Form Freeze-dried
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.