Learn More
ZeptoMetrix NATtrol™ Flu/RSV/SARS-CoV-2 Swab Control Kit (4 swabs)
SDP
Catalog No. 22046610 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Detectable Analytes:
Influenza A, COVID-19, RSV, Influenza B
Catalog No. Detectable Analytes
22-046-610 Influenza A, COVID-19, RSV, Influenza B
1 options
Catalog No. 22-046-610 Supplier ZeptoMetrix Supplier No. NATFRCKLSWIVD
Add to Cart
Add to Cart
Kit contains both Positive Control Swabs and Negative Control Swabs for comprehensive assay validation.
  • Positive Control Swab contains purified, intact viral particles that have been chemically modified to render them non-infectious and human A-549 cells
  • Each positive swab contains Influenza A (Brisbane/10/07), Influenza B (Florida/02/06), RSV B (CH93(18)-18), SARS-CoV-2 (USA-WA1/2020)1 and A-549 cells
  • The analytes are formulated in a proprietary matrix and dried on swabs
  • Negative Control Swab contains human A-549 cells formulated in a proprietary matrix and dried on swabs
  • Positive and Negative Control Swabs are full process controls designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures
Specifications
Detectable Analytes Influenza A, COVID-19, RSV, Influenza B
Type Control Kit
Form Liquid