Description
- External run control intended to be used with molecular assays that detect nucleic acids from Chlamydia trachomatis (LGVII434), Neisseria gonorrhoeae, Trichomonas vaginalis and Mycoplasma genitalium (SEA-1)
- The analytes are formulated in a proprietary matrix and dried on swabs
- The control can be used to monitor the process steps of extraction, amplification and detection
- The control can also be used for training and evaluation of operator variation
Specifications
Specifications
| Format | Vial |
| Product Type | Quality Control |
| Sterility | Non-sterile |
| Fill Volume | 0.2 mL |
| No. per Pack | 24 Vials |