Learn More
ZeptoMetrix NATtrol Gastrointestinal Panels Verification Kit (15 x 0.2 mL)
Shop All ZeptoMetrix Corporation Products
Description
- Can be used to develop procedures for verification of performance of molecular assays that detect the presence of nucleic acids from Adenovirus F40/41, Astrovirus, Blastocystis spp., Campylobacter spp., Clostridioides difficile (tcdA/tcdB), Cryptosporidium spp., Cyclospora cayetanensis, Dientamoeba fragilis, Entamoeba histolytica, Enterotoxigenic E. coli (ETEC) LT/ST, Giardia lamblia, Microsporidia, Norovirus GI/GII, Plesiomonas shigelloides, Rotavirus A, Salmonella spp., Sapovirus I/II/IV/V, Shiga-like toxin-producing E. coli (STEC) stx1, Shiga-like toxin-producing E. coli (STEC) stx2, Shigella/Enteroinvasive E. coli (EIEC), Strongyloides stercoralis, Vibrio spp., Vibrio cholerae, and Yersinia enterocolitica
- The panel can also be used for training and evaluation of operator proficiency
- The positive controls contain purified, intact microorganisms and viruses that have been chemically modified to render them non-infectious and refrigerator stable and are formulated in a proprietary matrix
- The negative control is formulated in the absence of any microorganisms or viruses and contains proprietary matrix only
Specifications
Specifications
| Format | Vial |
| Product Type | Verification Panel |
| Sterility | Non-sterile |
| Fill Volume | 0.2 mL |
| No. per Pack | 15 Vials |