Learn More
ZeptoMetrix NATtrol™ Group A Strep Positive Control
SDP
Catalog No. 22158263 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.25 mL
Catalog No. Quantity
22-158-263 6 x 0.25 mL
1 options
Catalog No. 22-158-263 Supplier ZeptoMetrix Supplier No. NATSPY6L
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Designed to evaluate the performance of nucleic acid tests for determination of the presence of viral and bacterial nucleic acids
Formulated with purified, intact virus particles and bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable.
Specifications
Detects Viral and bacterial nucleic acids
For Use With (Application) Verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 6 x 0.25 mL