Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Group A Streptococcus (GAS) Positive Control Swab (4 Swabs)
SDP

Catalog No. 22021870 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
4/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-021-870 4/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-021-870 Supplier ZeptoMetrix Supplier No. NATGASSW
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains purified, intact Streptococcus pyogenes that have been chemically modified to render them non-infectious.

  • It is an external run control intended to be used with molecular assays that detect nucleic acids from Streptococcus pyogenes
  • The control can be used to monitor the process steps of extraction, amplification, and detection
  • The control can also be used for training and evaluation of operator variation
  • The control is formulated in a proprietary matrix and dried on swabs

Specifications

CE Marker No
Clia Complexity High
Detectable Analytes Streptococci (Group A)
Type QC
Quantity 4/Pk.
Form Swab
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.