Learn More
ZeptoMetrix NATtrol™ HPV 16 Stock
SDP
Catalog No. 21200375 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
21-200-375 Non-sterile
1 options
Catalog No. 21-200-375 Supplier ZeptoMetrix Supplier No. NATHPV16STQ
Add to Cart
Add to Cart
NATtrol™ HPV 16 Stock (NATHPV16-STQ) contains CaSki cells that contain human papillomavirus type 16 (HPV16) DNA integrated into the genome
  • The cells have been chemically modified to render them refrigerator stable.
  • The control is supplied in a proprietary matrix.
Specifications
Product Type HPV 16 Stock
Sterility Non-sterile
Quantity 1 mL