Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Human Hepatitis B
SDP

Catalog No. 22157786 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
10000IU/mL
400IU/mL
50000IU/mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-157-786 10000IU/mL
22-157-785 50000IU/mL
22-157-783 400IU/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 22-157-786 Supplier ZeptoMetrix Supplier No. NATHBV0006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal control for amplification assays

  • Formulated from highly purified infectious viruses, from cell or plasma of infected individuals
  • After purification virus is chemically and enzymatically modified to alter surface proteins
  • Provide completely intact viral particle that is unable to bind, penetrate or infect any host cell

Human Source Material used in the manufacture of this product has been tested and found to be negative for HBsAg, antibodies to HIV-1/2 and HCV. Handle as if capable of transmitting infectious agents; use Universal Precautions when working with this product.

Used as a control/calibrators for PCR, TMA, Branched Chain DNA and NAT assays

Specifications

Certifications/Compliance ISO, 13485
Concentration 10000IU/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Virus Stock
Formulation Formulated with purified, intact virus particles
For Use With (Application) Determination of presence of HBV DNA, use as controls and calibrators for PCR, TMA, Branched Chain DNA and other NAT assays
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.