Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Human Metapneumovirus Peru6-2003 Stock
SDP

Catalog No. 22156741 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-741 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-156-741 Supplier ZeptoMetrix Supplier No. NATHMPVST
Add to Cart
Add to Cart

For determination of the presence of HMPV RNA

  • Concentration range from 22 to 25
  • Strain is Peru6-2003
  • 1.0mL vial
  • Formulated with purified, intact virus particles

IMPORTANT: Contains Metapneumovirus. Before vendor can ship order, customer must submit a Certificate of Compliance (COC) regarding use restrictions.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Certifications/Compliance ISO, 13485
Concentration 22 to 25
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Virus Stock
For Use With (Application) Determination of presence of HMPV RNA
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.