Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Staphylococcus aureus (MRSA) COL External Run Controls
SDP

Catalog No. 22156904 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Concentration:
50000cfu/mL
5000cfu/mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
22-156-904 50000cfu/mL
22-156-759 5000cfu/mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-156-904 Supplier ZeptoMetrix Supplier No. NATSAU(MRSA)ERCM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Formulated with purified, intact chemically modified bacterial cells to render them non-infectious and refrigerator stable

  • Contains 6 x 1.0mL vials of S. aureus (MRSA) NATtrol™ at target concentrations of 5,000 to 50,000cfu/mL
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate the performance of nucleic acid tests for determination of the presence of S. aureus DNA

Contain intact virus and should be run in a manner identical to that used for clinical specimens.

Validation of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Biosafety Level 2
Certifications/Compliance ISO, 13485
Concentration 50000cfu/mL
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type External Run Control
Formulation Formulated with purified, intact bacterial cells
For Use With (Application) Determination of presence of S. aureus DNA, validation of clinical assays, development of diagnostic tests and training of laboratory personnel
Quantity 6 x 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.