Learn More
ZeptoMetrix NATtrol T.vaginalis Negative Control
SDP
Catalog No. 22158042 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 1.2 mL
Catalog No. Quantity
22-158-042 6 x 1.2 mL
1 options
Catalog No. 22-158-042 Supplier ZeptoMetrix Supplier No. NATTVNEG6MC
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid test for determination of the presence of T. vaginalis nucleic acids
  • Formulated with purified, intact organisms that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
Specifications
Quantity 6 x 1.2 mL