Description
- The target concentration for each gene segment is 1.0E+06 copies/ mL, measured by digital PCR.
- The phage-like particles have been chemically modified to render them refrigerator stable. The control is supplied in a proprietary matrix.
Specifications
Specifications
| Product Type | Influenza A H5N1 Quantitative Stock |
| Sterility | Non-sterile |
| Quantity | 1 X 1.0 mL |