Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Influenza/RSV Negative Control
SDP

Catalog No. 22158212 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 X 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-212 6 X 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-212 Supplier ZeptoMetrix Supplier No. NATCXVA96CIVD
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use in evaluating and monitoring of qualitative molecular diagnostic assays for the detection of the absence of Influenza/RSV nucleic acid.

  • Formulated in a proprietary purified protein matrix that mimics the composition of a true clinical specimen.Formulated in a Purified Protein Matrix that is fully commutable with true clinical specimens
  • Can be used for verification of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Evaluating and monitoring of qualitative molecular diagnostic assays for the detection of the absence of Influenza/RSV nucleic acid.
Includes 6 x 0.5 mL vials of NATtrol™ Coxsackievirus formulated in a Purified Protein Matrix that is fully commutable with true clinical specimens.
Quantity 6 X 0.5 mL

This product is an IVD/CE marked USA FDA Class 1 exempt, unassayed external run control intended for in vitro diagnostic use and are for professional use only. NATtrol™  controls are not intended for use as a substitute for the internal controls provided by in vitro diagnostic kit manufacturers. Quality control materials should be used in accordance with local, state, federal and accreditation requirements.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.