Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Influenza Verification Panel
SDP

Catalog No. 22158175 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
18 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-175 18 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-175 Supplier ZeptoMetrix Supplier No. NATFVP(XP)C
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to evaluate the performance of nucleic acid tests for determination of the presence of viral nucleic acids. Can also be used for verification of clinical assays, development of diagnostic tests and training of laboratory personnel.

Panel Contains:
  • 3 tubes each of the Influenza A H1 and Influenza A H3 positive members
  • 6 tubes of the Influenza B positive member
  • 6 tubes of Coxsackievirus A9 negative member

The panel members are supplied in a purified protein matrix that mimics the composition of a true clinical specimen.

Panel Members:
  • Influenza A H1 A; Strain: /New Caledonia/20/99; Expected Result: Flu A POSITIVE, Flu B NEGATIVE
  • Influenza A H3; Strain: A/Brisbane/10/07; Expected Result: Flu A POSITIVE, Flu B NEGATIVE
  • Influenza B; Strain: B/Florida/02/06; Expected Result: Flu A NEGATIVE, Flu B POSITIVE
  • Coxsackievirus Type A9; Strain: NA; Expected Result: Flu A NEGATIVE,Flu B NEGATIVE

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) Evaluate the performance of nucleic acid tests for determination of the presence of viral nucleic acids.
Quantity 18 x 0.5 mL
Regulatory Status RUO

For Research Use Only. Not for use in humans. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.