Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Meningitis/Encephalitis (ME) Controls
SDP

Catalog No. 22158048 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.2mL vials of ME Control 1 and 6 x 0.2mL vials of ME Control 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-048 6 x 0.2mL vials of ME Control 1 and 6 x 0.2mL vials of ME Control 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-048 Supplier ZeptoMetrix Supplier No. NATMECBIO
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Each control contains 6 x 0.2mL vials of ME Control 1 and 6 x 0.2mL vials of ME Control 2
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate performance of nucleic acid tests for determination of the presence of viral nucleic acids
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimen

Specifications

Content And Storage 2°C to 8°C
Product Type Meningitis/Encephalitis (ME) Controls
Quantity 6 x 0.2mL vials of ME Control 1 and 6 x 0.2mL vials of ME Control 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.