Learn More
ZeptoMetrix™ NATtrol Mpox Virus Positive Control (6 x 0.5 mL)
SDP
Catalog No. 22158473 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
Catalog No. Quantity
22-158-473 6 x 0.5 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-158-473 Supplier ZeptoMetrix™ Supplier No. NATMPXVPOS6CIVD
Add to Cart
Add to Cart
This item is not returnable. View return policy
NATtrol™ Mpox Virus Positive Control contains purified, intact Mpox viral particles that have been chemically modified to render them non-infectious and refrigerator stable.
  • Contains purified, intact Mpox viral particles that have been chemically modified to render them non-infectious and refrigerator stable
  • Formulated in a proprietary matrix
  • A full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures
Specifications
Product Type Positive Control
Sterility Non-sterile
Quantity 6 x 0.5 mL