Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ MRSA Panel
SDP

Catalog No. 21100551 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
4 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-100-551 4 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-100-551 Supplier ZeptoMetrix Supplier No. NATMRSAPC
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to evaluate performance of nucleic acid tests for determination of the presence of S. aureus DNA

  • Contains 4 x 0.5mL
  • Formulated with purified, intact chemically modified bacterial particles to render them non-infectious and refrigerator stable
  • Supplied in purified protein matrix that mimics the composition of a true clinical specimen
Panel Members:
  • S. aureus (MRSA Community Strain)
  • S. aureus (MRSA Hospital Strain)
  • S. aureus (MSSA)
  • S. epidermidis (MSSE HER 1292)
  • For international customers only (Requires a license authorizing export from the U.S. prior to shipment)

    For verification of clinical assays, development of diagnostic testsand training of laboratory personnel.

Specifications

Certifications/Compliance ISO, 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type NATtrol Verification Panel
For Use With (Application) For evaluating Nucleic Acid tests for S. aureus DNA
Quantity 4 x 0.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.