Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ MRSA Positive Control
SDP

Catalog No. 22158317 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.35 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-317 6 x 0.35 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-317 Supplier ZeptoMetrix Supplier No. NATMRSA6L

missing translation for 'validMainframeMsg'

NATtrol™ MRSA Positive Control is formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable.

  • NATtrol products are ready to use, inactivated full process controls designed to evaluate performance of molecular tests. They can be used for verification of assays, training of laboratory personnel and to monitor assay-kit lot performance.
  • NATtrol products contain intact organisms and should be run in a manner similar to clinical specimens.
  • Quality Control

Specifications

Content And Storage Positive Control should be stored at 2 to 8°C
Description Contains inactivated bacterial cells. Should be handled as if potentially infectious.
Product Type Culture Fluid
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Includes Each control pack contains 6 x 0.35 mL vials of NATtrol™ S. aureus (MRSA). These controls are supplied in a purified protein matrix that mimics the composition of a true clinical specimen.
Quantity 6 x 0.35 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.