Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Neisseria gonorrhoeae Positive Control Pack
SDP

Catalog No. 22156113 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 1.25 mL Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-156-113 6 x 1.25 mL Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-156-113 Supplier ZeptoMetrix Supplier No. NATNG6MCIVD
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable

  • Each control contains 6 x 1.25mL vials of NATtrol Neisseria gonorrhoeae formulated in a Purified Protein Matrix
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate performance of nucleic acid tests for determination of presence of respiratory virus nucleic acids
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimens

Specifications

Content And Storage 2°C to 8°C
Product Type Rapid Test Control Pack
For Use With (Application) Quality control of clinical assays and training of laboratory personnel
Quantity 6 x 1.25 mL Vials

This product is an IVD/CE marked USA FDA Class 1 exempt, unassayed external run control intended for in vitro diagnostic use and are for professional use only. NATtrol™ controls are not intended for use as a substitute for the internal controls provided by in vitro diagnostic kit manufacturers. Quality control materials should be used in accordance with local, state, federal and accreditation requirements.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.