Learn More
Zeptometrix NATtrol™ Respiratory Panel 2.1 (RP2.1) Controls
SDP
Catalog No. 22158415 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
12 x 0.3 mL
Catalog No. Quantity
22-158-415 12 x 0.3 mL
1 options
Catalog No. 22-158-415 Supplier Zeptometrix Supplier No. NATRPC2.1BIO
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial and viral nucleic acids.
NATtrol™ Respiratory Panel 2.1 (RP2.1) Controls (qualitative) is formulated with purified, intact bacterial cells and viral particles. The microorganisms have been chemically modified to render them non-infectious and refrigerator stable.
NATRPC2.1-BIO contains 6 x 0.3 mL vials of RP2.1 Control 1 and 6 x 0.3 mL of RP2.1 Control 2. The controls are supplied in a proprietary matrix.
INTENDED USE:
  • NATtrol™ RP2.1 Controls are designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial and viral nucleic acids.
  • NATtrol™ Respiratory Panel 2.1 (RP2.1) Controls can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel.
Specifications
Type Controls
For Use With (Application) Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 12 x 0.3 mL
RUO only.