Learn More
ZeptoMetrix™ NATtrol Respiratory Panel Control
SDP
Catalog No. 23900533 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.35 mL
Catalog No. Quantity
23-900-533 6 x 0.35 mL
1 options
Catalog No. 23-900-533 Supplier ZeptoMetrix™ Supplier No. NATRSPCDIA
Add to Cart
Add to Cart
A qualitative external control intended to be used with molecular assays that detect nucleic acids from the organisms.
NATtrol™ Respiratory Panel Control contains purified, intact bacteria and viruses that have been chemically modified to render them non-infectious and refrigerator stable. The control is supplied in a proprietary matrix.
Specifications
CE Marker No
Clia Complexity Not Applicable
Detectable Analytes Nucleic Acid
Quantity 6 x 0.35 mL