Learn More
ZeptoMetrix NATtrol Respiratory/Sore Throat (R/ST) Verification Panel
SDP
Catalog No. 21124404 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
25 x 0.6 mL, 6 x 1.8 mL
Catalog No. Quantity
21-124-404 25 x 0.6 mL, 6 x 1.8 mL
1 options
Catalog No. 21-124-404 Supplier ZeptoMetrix Supplier No. NATRSTBIO
Add to Cart
Add to Cart
A diagnostic that can be used to develop procedures for verification of performance of molecular assays that detect the presence of bacterial and viral nucleic acids.
  • Contains purified, intact bacteria and viruses that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a proprietary matrix.
Specifications
Product Type Verification Panel
Sterility Non-sterile
Quantity 25 x 0.6 mL, 6 x 1.8 mL