Learn More
Zeptometrix NATtrol™ Respiratory Verification Panel 2.1
SDP
Catalog No. 22158408 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
24 x 0.6 mL
Catalog No. Quantity
22-158-408 24 x 0.6 mL
1 options
Catalog No. 22-158-408 Supplier Zeptometrix Supplier No. NATRVP2.1BIO
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial and viral nucleic acids.
NATtrol™ Respiratory Verification Panel 2.1 (qualitative) is formulated with purified, intact bacterial cells and viral particles. The microorganisms have been chemically modified to render them non-infectious and refrigerator stable. NATRVP2.1-BIO contains 23 x 0.6 mL vials of bacterial and viral NATtrol™ and 1 x 0.6 mL of negative (matrix only). The panel members are supplied in a proprietary matrix.
INTENDED USE:
  • NATtrol™ Respiratory Verification Panel 2.1 is designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial and viral nucleic acids.
  • NATRVP2.1-BIO enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Certifications/Compliance ISO 13485
Product Type Panel
For Use With (Application) Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 24 x 0.6 mL
Regulatory Status RUO
RUO only.