Learn More
Zeptometrix NATtrol™ Respiratory Verification Panel 2
SDP
Catalog No. 22158416 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
21 x 0.25 mL
Catalog No. Quantity
22-158-416 21 x 0.25 mL
1 options
Catalog No. 22-158-416 Supplier Zeptometrix Supplier No. NATRVP2QIA
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial, fungal, and nucleic acids.
NATtrol™ Respiratory Verification Panel 2 (qualitative) is formulated with purified, intact bacterial/fungal cells and viral particles that have been chemically modified to render them non-infectious and refrigerator stable. NATRVP2-QIA contains 20 x 0.25 mL vials of bacterial, fungal, or viral NATtrol™ and 1 x 0.25 mL vial of negative (matrix only). The panel members are supplied in a proprietary matrix.
INTENDED USE:
  • NATtrol™ Respiratory Verification Panel 2 is designed to evaluate the performance of nucleic acid tests for determination of the presence of bacterial, fungal, and nucleic acids.
  • NATRVP2- QIA can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel.
Specifications
Certifications/Compliance ISO 13485
Product Type Panel
For Use With (Application) Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 21 x 0.25 mL
Regulatory Status RUO
RUO only.