Learn More
ZeptoMetrix™ NATtrol Respiratory Verification Panel
SDP
Catalog No. 23900534 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.35 mL
Catalog No. Quantity
23-900-534 6 x 0.35 mL
1 options
Catalog No. 23-900-534 Supplier ZeptoMetrix™ Supplier No. NATRSPDIA
Add to Cart
Add to Cart
Can be used to develop procedures for verification of performance of molecular assays that detect the presence of bacterial and viral nucleic acids from the organisms.
NATtrol™ Respiratory Verification Panel contains purified, intact bacteria and viruses that have been chemically modified to render them non-infectious and refrigerator stable. The panel members are supplied in a proprietary matrix.
Specifications
Product Type Verification Panel
Sterility Non-sterile
Quantity 6 x 0.35 mL