Learn More
ZeptoMetrix NATtrol™ Respiratory Verification Panel
SDP
Catalog No. 22158045 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
15 x 0.5 mL vials of bacterial/viral targets, 2 x 1 mL vials of Negative Control
Catalog No. Quantity
22-158-045 15 x 0.5 mL vials of bacterial/viral targets, 2 x 1 mL vials of Negative Control
1 options
Catalog No. 22-158-045 Supplier ZeptoMetrix Supplier No. NATRVPNNS
Add to Cart
Add to Cart
Formulated with purified, intact chemically modified virus particles to render them non-infectious and refrigerator stable
  • Each control contains 15 x 0.5mL vials of bacterial/viral targets, 2 x 1mL vials of Negative Control
  • Supplied in purified protein matrix identical to the composition of true clinical specimen
  • Designed to evaluate performance of nucleic acid tests for determination of the presence of viral nucleic acids
  • Contain intact organisms and should be run in a manner identical to that used for clinical specimen
Specifications
Content And Storage 2°C to 8°C
Product Type Respiratory Verification Panel
Quantity 15 x 0.5 mL vials of bacterial/viral targets, 2 x 1 mL vials of Negative Control