Learn More
Zeptometrix NATtrol™ S. aureus (MRSA) Stock (Quantitative)
SDP
Catalog No. 22158410 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
22-158-410 1 mL
1 options
Catalog No. 22-158-410 Supplier Zeptometrix Supplier No. NATSAUSTQ
Add to Cart
Add to Cart
Designed to evaluate the performance of nucleic acid tests for determination of the presence of S. aureus (MRSA) nucleic acid.
NATtrol™ S. aureus (MRSA) Stock is formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable. Each vial contains 1.0 mL of NATtrol™ S. aureus (MRSA) stock in a proprietary matrix with a target concentration.
INTENDED USE:
  • NATtrol™ S. aureus (MRSA) stock is designed to evaluate the performance of nucleic acid tests for determination of the presence of S. aureus (MRSA) nucleic acid.
  • NATtrol™ S. aureus (MRSA) stock enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Certifications/Compliance ISO 13485
Concentration 1,000,000 cp/mL
Product Type NATtrol Virus Stock
For Use With (Application) Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance
Quantity 1 mL
Regulatory Status RUO
RUO only.