Learn More
Description
NATtrol™ S. pneumoniae Stock is formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable. Each vial contains 1.0 mL of NATtrol™ S. pneumoniae stock in a proprietary matrix with a target concentration.
INTENDED USE:
- S. pneumoniae stock is designed to evaluate the performance of nucleic acid tests for determination of the presence of S. pneumoniae nucleic acid.
- NATtrol™ S. pneumoniae stock enables laboratories to monitor test variation, lot-to-lot test kit performance, operator variation, and can provide assistance in identifying random or systemic error.
Specifications
Specifications
| Certifications/Compliance | ISO 13485 |
| Concentration | 1,000,000 cp/mL |
| Product Type | NATtrol Virus Stock |
| For Use With (Application) | Evaluate performance of molecular tests, verification of assays, training of laboratory personnel and to monitor assay-kit lot performance |
| Quantity | 1 mL |
| Regulatory Status | RUO |
RUO only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
