Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ SARS-CoV-2 (E/ORF1ab recombinant) Stock
SDP

Catalog No. 22158342 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-342 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-342 Supplier ZeptoMetrix Supplier No. 0831043
 Restricted Item
Add to Cart
Add to Cart

NATtrol™ SARS-CoV-2 (E/ORF1ab recombinant) Stock is formulated with purified, intact bacterial cells containing synthetic SARS-CoV-2 sequence.

  • Designed to evaluate the performance of nucleic acid tests for determination of the presence of SARS-CoV-2 E gene and portions of the ORF1ab gene region.
  • Can also be used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel.
  • The cells have been chemically modified to render them noninfectious and refrigerator stable.
  • Although it contains inactivated bacteria, it should be handled as if potentially infectious.

Specifications

Content And Storage Stock should be stored at 2°C to 8°C
Concentration 22 to 25 Ct Range
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Includes Each vial contains 1.0 mL of NATtrol™ SARS-CoV-2 (E/ORF1ab recombinant) Stock in a purified protein matrix.
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.