Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ SARS-CoV-2 (recombinant) Stock
SDP

Catalog No. 22158340 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-340 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-340 Supplier ZeptoMetrix Supplier No. 0831042
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

NATtrol™ SARS-CoV-2 (recombinant) Stock is formulated with purified, intact bacterial cells that have been chemically modified to render them non-infectious and refrigerator stable.

  • NATtrol products are ready to use, inactivated full process controls designed to evaluate performance of molecular tests. They can be used for verification of assays, training of laboratory personnel and to monitor assay-kit lot performance.
  • NATtrol products contain intact organisms and should be run in a manner similar to clinical specimens.
  • NATtrol SARS-CoV-2 (recombinant) Stock is formulated with purified, intact bacterial cells containing synthetic SARS-CoV-2 sequence.
  • Standard/Control

Specifications

Content And Storage Stock should be stored at 2°C to 8°C
Concentration 22 to 25 Ct Range
For Use With (Equipment) To avoid cross-contamination, use separate pipette tips for all reagents.
Includes Each vial contains 1.0 mL of NATtrol™ SARS-CoV-2 (recombinant) Stock in a purified protein matrix
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.