Learn More
ZeptoMetrix NATtrol™ Strep A Positive Control
SDP
Catalog No. 21403165 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
6 x 0.5 mL
Catalog No. Quantity
21-403-165 6 x 0.5 mL
1 options
Catalog No. 21-403-165 Supplier ZeptoMetrix Supplier No. NATSPY6MCIVD
Add to Cart
Add to Cart
NATtrol™ Strep A Positive Control contains purified, intact Streptococcus pyogenes that have been chemically modified to render them non-infectious and refrigerator stable.
  • The control is formulated in a proprietary matrix.
  • NATtrol™ Strep A Positive Control is a full process control designed to monitor the steps of extraction, amplification, and detection in nucleic acid testing procedures.
Specifications
CE Marker No
Clia Complexity Not Applicable
Detectable Analytes Streptococci (Group A)
Type Positive Control
Quantity 6 x 0.5 mL
Form Liquid