Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Vaginal Panel
SDP

Catalog No. 22158104 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
24 x 0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-104 24 x 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-158-104 Supplier ZeptoMetrix Supplier No. NATVPBD
Add to Cart
Add to Cart
This item is not returnable. View return policy

Designed to evaluate the performance of nucleic acid tests for the determination of the presence of BV DNA markers that represent positive or negative BV depending on marker combination used, Candida species, and Trichomonas vaginalis DNA

NATtrol Vaginal Panel (NATVP-BD) panel contains 24 (4 of each panel member) x 0.5mL vials each containing NATtrol bacterial vaginosis markers, either Positive or Negative marker combination, and/or Candida species, and/or Trichomonas vaginalis

  • Formulated with purified, intact bacterial and fungal cells that have been chemically modified to render them non-infectious and refrigerator stable
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Can be used for verification of clinical assays, development of diagnostic tests and training of laboratory personnel

Specifications

Content And Storage 2°C to 8°C
Quantity 24 x 0.5 mL

Do not use in humans. For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.