Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix NATtrol™ Zika Virus Stock
SDP

Catalog No. 22158054 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-054 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-158-054 Supplier ZeptoMetrix Supplier No. NATZIKVST
Only null left
Add to Cart
Add to Cart

Designed to evaluate the performance of nucleic acid tests. ZeptoMetrix NATtrol™ Zika Virus Stock is used for validation of clinical assays, development of diagnostic tests and training of laboratory personnel.

  • Evaluates the performance of nucleic acid tests to determine the presence of Zika virus RNA
  • Formulated with purified, intact virus particles that have been chemically modified to render them non-infectious
  • Supplied in a purified protein matrix that mimics the composition of a true clinical specimen
  • Zika Virus Stock: (Conc.: 22-25)

Specifications

Certifications/Compliance ISO 13485
Content And Storage 2°C to 8°C
Form Solution
Media Type Vial
Product Type Zika Virus Stock
For Use With (Application) Determination of the presence of zika virus RNA
Quantity 1 mL

For research use only. Not for use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.