Learn More
MSC Seventh Generation Automatic Dishwasher Gel, Fresh Citrus, 65 oz, 6-count
Catalog No. 502836002
Encompass
This automatic dishwasher gel effectively cuts through grease, starch, and protein-based food residues using liquid detergent enzymes and surfactants. It's even effective on tough egg stains. The product is from a Certified B Corporation committed to environmental and social responsibility.
  • Cuts through grease and food with liquid detergent enzymes and surfactants.
  • Features PlanTech™ grease cutting technology for cleaner dishes.
  • Formulated with natural ingredients and plant-derived surfactants.
  • Effectively cleans ceramics, enamel, fiberglass, glass, metal, and plastic.
  • Tackles cooked-on grease, food particles, oils, and soil.
  • Provides a fresh citrus scent.
  • Non-toxic and VOC-free.
  • Meets Kosher, Leaping Bunny™, and USDA Certified Biobased 95% standards.
  • Comes in 65 oz bottles, with 6 per carton.
Catalog No. 50-283-6002 Supplier MSC Supplier No. 38702122
This automatic dishwasher gel effectively cuts through grease, starch, and protein-based food residues using liquid detergent enzymes and surfactants. It's even effective on tough egg stains. The product is from a Certified B Corporation committed to environmental and social responsibility.
  • Cuts through grease and food with liquid detergent enzymes and surfactants.
  • Features PlanTech™ grease cutting technology for cleaner dishes.
  • Formulated with natural ingredients and plant-derived surfactants.
  • Effectively cleans ceramics, enamel, fiberglass, glass, metal, and plastic.
  • Tackles cooked-on grease, food particles, oils, and soil.
  • Provides a fresh citrus scent.
  • Non-toxic and VOC-free.
  • Meets Kosher, Leaping Bunny™, and USDA Certified Biobased 95% standards.
  • Comes in 65 oz bottles, with 6 per carton.