Learn More
Qorpak™ Natural HDPE Wide Mouth Round Bottle
Description
- Ideal for general lab use with solids and powders
- These HDPE (high-density polyethylene) bottles provide an excellent moisture barrier, are translucent, and are impact resistant
- These bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping
Specifications
Specifications
| Capacity (Metric) | 240 mL |
| Color | Natural |
| Material | HDPE |
| No. per Case | 250 |
| Packaging | Bulk |
| Sterility | Non-sterile |
| Neck Finish | 48-400 |
| Closure Included | No |
| Graduated | No |
| Mouth | Wide, Threaded |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.