Learn More
Qorpak™ Natural LDPE Narrow Mouth Nasal Spray/Dropper Bottles with Caps
Description
- Bottles, spray plugs, and overcaps are bagged then placed in box
- Components arrive unassembled
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| CAS | 67-56-1 |
| Color | Natural |
| Concentration | 100 μg/mL in methanol (as free base) |
| Packaging | Ampule |
| Includes | Bottle, Spray/Dropper Tip, Overcap |
| Material | LDPE |
| Linear Formula | C18H19D3INO3 · HCl |
| MDL Number | MFCD18452514 |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.